ADV390217839 AChemBlock

1H-Pyrrolo[3,2-b]pyridine 4-oxide

  • MDL number MFCD12827548
  • Cas Number : 1116136-36-7
  • Molecular Weight : 134.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/1h-pyrrolo-3-2-b-pyridine-4-oxide.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6356
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart