ADV390217840 AChemBlock

2,8-Dichloro-1,5-naphthyridine

  • MDL number MFCD20926182
  • Cas Number : 28252-76-8
  • Molecular Weight : 199.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-8-dichloro-1-5-naphthyridine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6358
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart