ADV390217843 AChemBlock

Ethyl 4-chloro-6-methyl-2-(methylthio)pyrimidine-5-carboxylate

  • MDL number MFCD08063161
  • Cas Number : 583878-42-6
  • Molecular Weight : 246.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form yellow liquid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/ethyl-4-chloro-6-methyl-2-methylthio-pyrimidine-5-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6361
pictograms GHS07
hstatements H302;H313;H319; H335
Availability details for
Quantity
  • Availability
  • Add to Cart