ADV390217859 AChemBlock

1-Benzyl-1,2,3,6-tetrahydropyridine

  • MDL number MFCD11501660
  • Cas Number : 40240-12-8
  • Molecular Weight : 173.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1-benzyl-1-2-3-6-tetrahydropyridine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6390
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart