ADV390217871 AChemBlock

(3aR,6aR)-3a,6a-dihydro-2,2-dimethyl-4H-cyclopenta-1,3-dioxol-4-one

  • MDL number MFCD08166479
  • Cas Number : 115509-13-2
  • Molecular Weight : 154.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form light-yellow solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3ar-6ar-3a-6a-dihydro-2-2-dimethyl-4h-cyclopenta-1-3-dioxol-4-one.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6405
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart