ADV390217872 AChemBlock

3-Fluoro-1-[tris(1-methylethyl)silyl]-1H-pyrrole

  • MDL number MFCD20926185
  • Cas Number : 156304-02-8
  • Molecular Weight : 241.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/3-fluoro-1-tris-1-methylethyl-silyl-1h-pyrrole.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6407
pictograms GHS07
hstatements H302;H313;H315;H335
Availability details for
Quantity
  • Availability
  • Add to Cart