ADV390217884 AChemBlock

1-Boc-4-aminomethyl-4-hydroxypiperidine

  • MDL number MFCD11579803
  • Cas Number : 392331-66-7
  • Molecular Weight : 230.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1-boc-4-aminomethyl-4-hydroxypiperidine.html

Safety Information

signalword Warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6424
pictograms GHS07
hstatements H302;H313;H319; H335
Availability details for
Quantity
  • Availability
  • Add to Cart