ADV390217927 AChemBlock

3-Oxocyclobutyl acetate

  • MDL number MFCD11976057
  • Cas Number : 63930-59-6
  • Molecular Weight : 128.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/3-oxocyclobutyl-acetate.html
Availability details for
Quantity
  • Availability
  • Add to Cart