ADV390217931 AChemBlock

3,3-Difluorocyclobutanol

  • MDL number MFCD18791191
  • Cas Number : 637031-88-0
  • Molecular Weight : 108.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form colorless liquid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-3-difluorocyclobutanol.html
Availability details for
Quantity
  • Availability
  • Add to Cart