ADV390217940 AChemBlock

cis-4-(Boc-amino)-3-fluoropiperidine

  • MDL number MFCD23105958
  • Cas Number : 1268521-83-0
  • Molecular Weight : 218.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/cis-4-boc-amino-3-fluoropiperidine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6535
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart