ADV390217957 AChemBlock

3-(Boc-amino)-6-chloro-2-formylpyridine

  • MDL number MFCD18452130
  • Cas Number : 1199557-04-4
  • Molecular Weight : 256.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-boc-amino-6-chloro-2-formylpyridine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6562
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart