ADV390217964 AChemBlock

(R)-3-(Cbz-amino)piperidine

  • MDL number MFCD04115318
  • Cas Number : 478646-32-1
  • Molecular Weight : 234.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/r-3-cbz-amino-piperidine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6570
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart