ADV390217966 AChemBlock

3-Amino-2-methyl-5-nitropyridine

  • MDL number MFCD08272071
  • Cas Number : 51984-61-3
  • Molecular Weight : 153.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-amino-2-methyl-5-nitropyridine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6572
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart