ADV390217990 AChemBlock

2-Bromomethyl-5,8-dioxaspiro[3.4]octane

  • MDL number MFCD23106219
  • Cas Number : 1057641-71-0
  • Molecular Weight : 207.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-bromomethyl-5-8-dioxaspiro-3-4-octane.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6603
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart