ADV390218001 AChemBlock

N-Ethyloxetan-3-amine

  • MDL number MFCD18849120
  • Cas Number : 1341989-73-8
  • Molecular Weight : 101.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/n-ethyloxetan-3-amine.html
Availability details for
Quantity
  • Availability
  • Add to Cart