ADV390218003 AChemBlock

1-(tert-Butoxycarbonyl)-2-(methoxycarbonyl)-piperidine-4-carboxylic acid

  • MDL number MFCD16040215
  • Cas Number : 1255666-29-5
  • Molecular Weight : 287.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1-tert-butoxycarbonyl-2-methoxycarbonyl-piperidine-4-carboxylic-acid.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6619
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart