ADV390218011 AChemBlock

trans-Benzyl 3-hydroxycyclobutylcarbamate

  • MDL number MFCD23106305
  • Cas Number : 937376-05-1
  • Molecular Weight : 221.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/trans-benzyl-3-hydroxycyclobutylcarbamate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6628
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart