ADV390218014 AChemBlock

3-(Bromomethyl)cyclobutanone

  • MDL number MFCD17170393
  • Cas Number : 463961-43-5
  • Molecular Weight : 163.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form colorless liquid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/3-bromomethyl-cyclobutanone.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6631
pictograms GHS07
hstatements H302;H313;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart