ADV390218019 AChemBlock

Thietan-3-amine hydrochloride

  • MDL number MFCD19982300
  • Cas Number : 128861-78-9
  • Molecular Weight : 125.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/thietan-3-amine-hydrochloride.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6637
pictograms GHS07
hstatements H302;H315;H335
Availability details for
Quantity
  • Availability
  • Add to Cart