ADV390218021 AChemBlock

Diphenyl(trifluoromethyl)sulfonium trifluoromethanesulfonate

  • MDL number MFCD12912173
  • Cas Number : 147531-11-1
  • Molecular Weight : 404.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form yellow solid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/diphenyl-trifluoromethyl-sulfonium-trifluoromethanesulfonate.html

Safety Information

signalword Warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6640
pictograms GHS07
hstatements H302;H313;H319; H335
Availability details for
Quantity
  • Availability
  • Add to Cart