ADV390218035 AChemBlock

3-Benzyloxycyclobutane-1,1-dicarboxylic acid diethyl ester

  • MDL number MFCD07700259
  • Cas Number : 54166-15-3
  • Molecular Weight : 306.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-benzyloxycyclobutane-1-1-dicarboxylic-acid-diethyl-ester.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6667
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart