ADV390218036 AChemBlock

1,1,1-Trichloro-4-ethoxy-but-3-en-2-one

  • MDL number MFCD11520438
  • Cas Number : 59938-07-7
  • Molecular Weight : 217.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form light yellow liquid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1-1-1-trichloro-4-ethoxy-but-3-en-2-one.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6672
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart