ADV390218042 AChemBlock

7-Aza-spiro[3.5]nonane hydrochloride

  • MDL number MFCD23105809
  • Cas Number : 1414885-16-7
  • Molecular Weight : 161.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/7-aza-spiro-3-5-nonane-hydrochloride.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6680
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart