ADV390218052 AChemBlock

(S)-Tetrahydrofuran-3-carbonitrile

  • MDL number MFCD20926189
  • Cas Number : 1363378-18-0
  • Molecular Weight : 97.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/s-tetrahydrofuran-3-carbonitrile.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6696
pictograms GHS07
hstatements H302;H313;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart