ADV390218083 AChemBlock

6-Bromopyrrolo[1,2-f][1,2,4]triazin-4(3H)-one

  • MDL number MFCD11973831
  • Cas Number : 888721-83-3
  • Molecular Weight : 214.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/6-bromopyrrolo-1-2-f-1-2-4-triazin-4-3h-one.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5649
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart