ADV390218084 AChemBlock

Ethyl 6-chloroimidazo[1,2-b]pyridazine-3-carboxylate

  • MDL number MFCD11975600
  • Cas Number : 1150566-27-0
  • Molecular Weight : 225.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/ethyl-6-chloroimidazo-1-2-b-pyridazine-3-carboxylate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5650
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart