ADV390218092 AChemBlock

cis-3-Fluoro-4-piperidinol hydrochloride

  • MDL number MFCD23703603
  • Cas Number : 955028-89-4
  • Molecular Weight : 155.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form White solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/cis-3-fluoro-4-piperidinol-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5703
pictograms GHS07
hstatements H302;H313;H319; H335
Availability details for
Quantity
  • Availability
  • Add to Cart