ADV390218096 AChemBlock

6-Fluoronicotinimidamide hydrochloride

  • MDL number MFCD23106374
  • Cas Number : 1419101-31-7
  • Molecular Weight : 175.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/6-fluoronicotinimidamide-hydrochloride.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5718
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart