ADV390218098 AChemBlock

(S)-tert-Butyl 2-(2-chloroacetyl)azetidine-1-carboxylate

  • MDL number MFCD11975857
  • Cas Number : 1260616-94-1
  • Molecular Weight : 233.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/s-tert-butyl-2-2-chloroacetyl-azetidine-1-carboxylate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5728
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart