ADV390218106 AChemBlock

4,5-O-isopropylidene-2-C-methyl-D-arabinonate

  • MDL number MFCD11112138
  • Cas Number : 93635-76-8
  • Molecular Weight : 248.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-5-o-isopropylidene-2-c-methyl-d-arabinonate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5757
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart