ADV390218112 AChemBlock

Imidazole-2-amidine acetate

  • MDL number MFCD20922762
  • Cas Number : 950685-49-1
  • Molecular Weight : 170.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/imidazole-2-amidine-acetate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5793
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart