ADV390218124 AChemBlock

6-Amino-4-fluoro-3-methyl-1H-indazole

  • MDL number MFCD20926148
  • Cas Number : 1363382-44-8
  • Molecular Weight : 165.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/6-amino-4-fluoro-3-methyl-1h-indazole.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5853
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart