ADV390218133 AChemBlock

1-Methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrole

  • MDL number MFCD13182200
  • Cas Number : 953040-54-5
  • Molecular Weight : 207.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1-methyl-3-4-4-5-5-tetramethyl-1-3-2-dioxaborolan-2-yl-1h-pyrrole.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5908
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart