ADV390218135 AChemBlock

1-Boc-4-(hydroxymethyl)-4-methyl-piperidine

  • MDL number MFCD08062516
  • Cas Number : 236406-21-6
  • Molecular Weight : 229.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1-boc-4-hydroxymethyl-4-methyl-piperidine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5941
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart