ADV390218142 AChemBlock

Benzylidene-bis(tricyclohexylphosphine)dichlororuthenium

  • MDL number MFCD01090946
  • Cas Number : 172222-30-9
  • Molecular Weight : 837

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form violet powder
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/benzylidene-bis-tricyclohexylphosphine-dichlororuthenium.html

Safety Information

signalword Warning
pcodes P210;P240;P241;P261;P264;P271;P280;P302+352;P304+340;P305+351+338;P337+313;P370+378;P403+233
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5956
pictograms GHS02; GHS07
hstatements H228;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart