ADV390218143 AChemBlock

Endo-3-amine-9-methyl-9-azabicyclo[3,3,1]nonane

  • MDL number MFCD07374273
  • Cas Number : 76272-56-5
  • Molecular Weight : 154.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form colorless or light-yellow liquid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/endo-3-amine-9-methyl-9-azabicyclo-3-3-1-nonane.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5962
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart