ADV390218156 AChemBlock

4-Hydroxy-5-methylpyrrolo[1,2-f][1,2,4]triazine

  • MDL number MFCD21647383
  • Cas Number : 529508-54-1
  • Molecular Weight : 149.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-hydroxy-5-methylpyrrolo-1-2-f-1-2-4-triazine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6062
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart