ADV390218161 AChemBlock

(R)-3-Hydroxymethylpyrrolidine hydrochloride

  • MDL number MFCD09800470
  • Cas Number : 1227157-98-3
  • Molecular Weight : 137.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form off-white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/r-3-hydroxymethylpyrrolidine-hydrochloride.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6083
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart