ADV390218172 AChemBlock

1-Boc-4-hydroxy-4-(hydroxymethyl)piperidine

  • MDL number MFCD08062514
  • Cas Number : 389889-80-9
  • Molecular Weight : 231.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1-boc-4-hydroxy-4-hydroxymethyl-piperidine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6161
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart