ADV390218175 AChemBlock

Methyl 6-bromoindole-4-carboxylate

  • MDL number MFCD07357290
  • Cas Number : 107650-22-6
  • Molecular Weight : 254.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form brown powder
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/methyl-6-bromoindole-4-carboxylate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6178
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart