ADV390218186 AChemBlock

N-(4-((6,7-Dimethoxyquinolin-4-yl)oxy)phenyl)-N-(4-fluorophenyl)cyclopropane-1,1-dicarboxamide

  • MDL number MFCD20926324
  • Cas Number : 849217-68-1
  • Molecular Weight : 501.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/n-4-6-7-dimethoxyquinolin-4-yl-oxy-phenyl-n-4-fluorophenyl-cyclopropane-1-1-dicarboxamide.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6215
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart