ADV390218194 AChemBlock

1-Benzyl-3,3-difluoro- 4,4-piperidinediol

  • MDL number MFCD20926176
  • Cas Number : 1067914-81-1
  • Molecular Weight : 243.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1-benzyl-3-3-difluoro-4-4-piperidinediol.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6277
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart