ADV390218198 AChemBlock

trans-tert-Butyl (3-(aminomethyl)cyclobutyl)carbamate

  • MDL number MFCD22123684
  • Cas Number : 1214727-57-7
  • Molecular Weight : 200.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/trans-tert-butyl-3-aminomethyl-cyclobutyl-carbamate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6303
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart