ADV390218200 AChemBlock

2-Phenyltetrahydrofuran

  • MDL number MFCD00961901
  • Cas Number : 16133-83-8
  • Molecular Weight : 148.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-phenyltetrahydrofuran.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5955
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart