ADV390218215 AChemBlock

Saxagliptin

  • MDL number MFCD12756398
  • Cas Number : 361442-04-8
  • Molecular Weight : 315.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Room Temp
Physical Form White solid
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/saxagliptin.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6374
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart