ADV390218217 AChemBlock

Alogliptin benzoate

  • MDL number MFCD09833195
  • Cas Number : 850649-62-6
  • Molecular Weight : 461.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Physical Form slight red powder
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/alogliptin-benzoate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6376
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart