ADV390218220 AChemBlock

Ethyl tetrahydropyranyl-4-acetate

  • MDL number MFCD03788560
  • Cas Number : 103260-44-2
  • Molecular Weight : 172.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form colorless liquid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/ethyl-tetrahydropyranyl-4-acetate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6386
pictograms GHS07
hstatements H302;H313;H315;H335
Availability details for
Quantity
  • Availability
  • Add to Cart