ADV390218255 AChemBlock

(4,4-Dimethyloxetan-2-yl)methanol

  • MDL number MFCD22123255
  • Cas Number : 61266-55-5
  • Molecular Weight : 116.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Physical Form colorless liquid
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/4-4-dimethyloxetan-2-yl-methanol.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6553
pictograms GHS07
hstatements H302;H313;H315;H335
Availability details for
Quantity
  • Availability
  • Add to Cart