ADV390218260 AChemBlock

1-(tetrahydrofuran-3-yl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole

  • MDL number MFCD18383269
  • Cas Number : 1029715-63-6
  • Molecular Weight : 264.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form slight yellow powder
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1-3-tetrahydrofuryl-1h-pyrazole-4-boronic-acid-pinacol-ester.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6573
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart