ADV390218268 AChemBlock

3-Bromo-1H-pyrrole-2-carboxylic acid methyl ester

  • MDL number MFCD23106341
  • Cas Number : 941714-57-4
  • Molecular Weight : 204.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-bromo-1h-pyrrole-2-carboxylic-acid-methyl-ester.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6702
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart